ENA245488018 Enamine

ethyl 2-[(2,2,2-trifluoroethyl)amino]acetate

  • MDL number MFCD11154791
  • Cas Number : 1021002-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55297
Availability details for
Quantity
  • Availability
  • Add to Cart