ENA245488026 Enamine

(4-cyanophenyl)methanesulfonohydrazide

  • MDL number MFCD11119785
  • Cas Number : 1039999-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55434
Availability details for
Quantity
  • Availability
  • Add to Cart