ENA245488027 Enamine

(3-cyanophenyl)methanesulfonohydrazide

  • MDL number MFCD12181286
  • Cas Number : 1156132-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55435
Availability details for
Quantity
  • Availability
  • Add to Cart