ENA245488031 Enamine

2-(3-acetylphenoxy)-N,N-dimethylacetamide

  • MDL number MFCD09949855
  • Cas Number : 1016886-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55441
Availability details for
Quantity
  • Availability
  • Add to Cart