ENA245488033 Enamine

2-tert-butylpyrimidin-5-amine

  • MDL number MFCD09753484
  • Cas Number : 59950-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55443
Availability details for
Quantity
  • Availability
  • Add to Cart