ENA245488042 Enamine

7-benzyl-1,3,7-triazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD12634012
  • Cas Number : 28937-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55451
Availability details for
Quantity
  • Availability
  • Add to Cart