ENA245488051 Enamine

N-(pyridin-3-yl)ethane-1-sulfonamide

  • MDL number MFCD03381351
  • Cas Number : 352707-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55307

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55307
Availability details for
Quantity
  • Availability
  • Add to Cart