ENA245488057 Enamine

2-(4-tert-butylphenyl)-2-oxoacetic acid

  • MDL number MFCD11103006
  • Cas Number : 7099-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55476

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-55476
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart