ENA245488059 Enamine

N-[(2,5-dimethoxyphenyl)methyl]-1-methyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD13196237
  • Cas Number : 1210939-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55479
Availability details for
Quantity
  • Availability
  • Add to Cart