ENA245488063 Enamine

5-methyl-1,3-dioxo-2-(2,2,2-trifluoroethyl)-2,3,3a,4,5,7a-hexahydro-1H-isoindole-4-carboxylic acid

  • MDL number MFCD13196209
  • Cas Number : 1212341-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55311
Availability details for
Quantity
  • Availability
  • Add to Cart