ENA245488065 Enamine

1-[(5-chlorothiophen-2-yl)methyl]piperidine-4-carboxylic acid

  • MDL number MFCD12169802
  • Cas Number : 1156269-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55489
Availability details for
Quantity
  • Availability
  • Add to Cart