ENA245488066 Enamine

N-(5-amino-2-fluorophenyl)-2-iodobenzamide

  • MDL number MFCD09042932
  • Cas Number : 926264-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55313
Availability details for
Quantity
  • Availability
  • Add to Cart