ENA245488069 Enamine

3-methoxy-4-(2-methoxyethoxy)benzoic acid

  • MDL number MFCD09810289
  • Cas Number : 247569-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55495
Availability details for
Quantity
  • Availability
  • Add to Cart