ENA245488076 Enamine

methyl 4-(4-bromophenyl)-2,4-dioxobutanoate

  • MDL number MFCD06740811
  • Cas Number : 60395-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55500
Availability details for
Quantity
  • Availability
  • Add to Cart