ENA245488090 Enamine

N-[1-(3-methoxyphenyl)ethyl]-1-methyl-1H-pyrazol-4-amine

  • MDL number MFCD12429441
  • Cas Number : 1153973-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55513
Availability details for
Quantity
  • Availability
  • Add to Cart