ENA245488097 Enamine

[(piperidin-3-yl)methyl](propan-2-yl)amine dihydrochloride

  • MDL number MFCD13196242
  • Cas Number : 1209558-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55582
Availability details for
Quantity
  • Availability
  • Add to Cart