ENA245488098 Enamine

N-[(2-methoxyphenyl)methyl]isoquinolin-5-amine

  • MDL number MFCD12648087
  • Cas Number : 1152972-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55583
Availability details for
Quantity
  • Availability
  • Add to Cart