ENA245488103 Enamine

1-(chloromethyl)-4-propylbenzene

  • MDL number MFCD11190944
  • Cas Number : 3166-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55594
Availability details for
Quantity
  • Availability
  • Add to Cart