ENA245488106 Enamine

1-methyl-N-(piperidin-4-yl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD11168086
  • Cas Number : 1152905-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55609
Availability details for
Quantity
  • Availability
  • Add to Cart