ENA245488112 Enamine

ethyl 2-{[5-(thiophen-2-yl)-1,3,4-oxadiazol-2-yl]sulfanyl}acetate

  • MDL number MFCD03118734
  • Cas Number : 459846-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55315
Availability details for
Quantity
  • Availability
  • Add to Cart