ENA245488128 Enamine

[6-(2-methoxyethoxy)-1,3-dioxaindan-5-yl]methanol

  • MDL number MFCD13196248
  • Cas Number : 1209106-01-3
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55642
Availability details for
Quantity
  • Availability
  • Add to Cart