ENA245488131 Enamine

4-(benzyloxy)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD06738564
  • Cas Number : 57928-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-202
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55645
Availability details for
Quantity
  • Availability
  • Add to Cart