ENA245488138 Enamine

2-(4-bromophenyl)-4-(chloromethyl)-1,3-oxazole

  • MDL number MFCD11200985
  • Cas Number : 22091-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55654

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55654
Availability details for
Quantity
  • Availability
  • Add to Cart