ENA245488139 Enamine

3-cyano-N-(prop-2-en-1-yl)benzamide

  • MDL number MFCD09940078
  • Cas Number : 1016832-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55321

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55321
Availability details for
Quantity
  • Availability
  • Add to Cart