ENA245488143 Enamine

N-butyl-2,2,6,6-tetramethylpiperidin-4-amine

  • MDL number MFCD00236393
  • Cas Number : 36177-92-1
  • Molecular Weight : 212.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55660
Availability details for
Quantity
  • Availability
  • Add to Cart