ENA245488146 Enamine

N-(cyclopentylmethyl)-1H-imidazole-1-carboxamide

  • MDL number MFCD12166736
  • Cas Number : 1096821-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55664
Availability details for
Quantity
  • Availability
  • Add to Cart