ENA245488149 Enamine

2-(2,3-dihydro-1H-inden-5-yl)-2-oxoacetic acid

  • MDL number MFCD11179655
  • Cas Number : 720707-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55323
Availability details for
Quantity
  • Availability
  • Add to Cart