ENA249152192 Enamine

1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carbaldehyde

  • MDL number MFCD13368189
  • Cas Number : 1221724-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51266
Availability details for
Quantity
  • Availability
  • Add to Cart