ENA249152207 Enamine

3-{[2-(2-chlorophenyl)-2-oxoethyl]carbamoyl}propanoic acid

  • MDL number MFCD13368207
  • Cas Number : 1221724-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-55910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55910
Availability details for
Quantity
  • Availability
  • Add to Cart