ENA249152212 Enamine

3-chloro-6-(propan-2-yloxy)pyridine-2-carbonitrile

  • MDL number MFCD13368212
  • Cas Number : 1221724-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56076
Availability details for
Quantity
  • Availability
  • Add to Cart