ENA249152213 Enamine

ethyl 2-(5-chloro-1-methyl-1H-imidazol-2-yl)-2-hydroxyacetate

  • MDL number MFCD13368213
  • Cas Number : 1221724-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56086
Availability details for
Quantity
  • Availability
  • Add to Cart