ENA249152224 Enamine

N-(5-methyl-1,2-oxazol-3-yl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD13368224
  • Cas Number : 1221724-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56196
Availability details for
Quantity
  • Availability
  • Add to Cart