ENA249152228 Enamine

ethyl 3-amino-2-benzylpropanoate hydrochloride

  • MDL number MFCD13368228
  • Cas Number : 103632-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56219
Availability details for
Quantity
  • Availability
  • Add to Cart