ENA249152229 Enamine

4-methyl-1-(1,2,3,4-tetrahydronaphthalen-1-yl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD13368229
  • Cas Number : 1235438-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56224
Availability details for
Quantity
  • Availability
  • Add to Cart