ENA249152230 Enamine

2-[(4-fluorophenyl)methyl]-3-hydroxypropanenitrile

  • MDL number MFCD13368230
  • Cas Number : 890147-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56228
Availability details for
Quantity
  • Availability
  • Add to Cart