ENA249152234 Enamine

methyl 5-formyl-3-methylthieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD13368234
  • Cas Number : 1221722-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56258
Availability details for
Quantity
  • Availability
  • Add to Cart