ENA249152239 Enamine

1-[4-(1H-1,2,4-triazol-1-yl)phenyl]methanamine dihydrochloride

  • MDL number MFCD13368241
  • Cas Number : 1221724-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56320
Availability details for
Quantity
  • Availability
  • Add to Cart