ENA249152242 Enamine

(8-bromo-2,3-dihydro-1,4-benzodioxin-6-yl)methanamine hydrochloride

  • MDL number MFCD13368244
  • Cas Number : 1221726-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233...235
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56353
Availability details for
Quantity
  • Availability
  • Add to Cart