ENA249152249 Enamine

trifluoromethanesulfonylethene

  • MDL number MFCD13368251
  • Cas Number : 18370-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56457
Availability details for
Quantity
  • Availability
  • Add to Cart