ENA249152251 Enamine

2-(4-methoxyphenyl)-1-(piperazin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD13368254
  • Cas Number : 1221723-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56484
Availability details for
Quantity
  • Availability
  • Add to Cart