ENA249152252 Enamine

2-(benzyloxy)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD13368256
  • Cas Number : 1235441-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56516

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-56516
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart