ENA249152258 Enamine

2-[(1H-imidazol-1-yl)methyl]aniline

  • MDL number MFCD09814189
  • Cas Number : 61292-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-55746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55746
Availability details for
Quantity
  • Availability
  • Add to Cart