ENA249152261 Enamine

2-{2-[(4-fluorophenyl)methyl]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD11205907
  • Cas Number : 1094423-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55785

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55785
Availability details for
Quantity
  • Availability
  • Add to Cart