ENA249152265 Enamine

4-methanesulfonamidobenzene-1-sulfonyl chloride

  • MDL number MFCD09048309
  • Cas Number : 93345-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51714
Availability details for
Quantity
  • Availability
  • Add to Cart