ENA249152268 Enamine

2-(methoxymethyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09258843
  • Cas Number : 1086380-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56033
Availability details for
Quantity
  • Availability
  • Add to Cart