ENA249152276 Enamine

N-(5-amino-2-methylphenyl)-2-iodobenzamide

  • MDL number MFCD09046311
  • Cas Number : 926208-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56072
Availability details for
Quantity
  • Availability
  • Add to Cart