ENA249152277 Enamine

3-(naphthalen-1-yl)-2,4-dioxo-1,2,3,4-tetrahydroquinazoline-7-carboxylic acid

  • MDL number MFCD09050997
  • Cas Number : 926268-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41158
Availability details for
Quantity
  • Availability
  • Add to Cart