ENA249152281 Enamine

1-{3-[2-(morpholin-4-yl)ethoxy]phenyl}methanamine

  • MDL number MFCD07772847
  • Cas Number : 857284-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56090
Availability details for
Quantity
  • Availability
  • Add to Cart