ENA249152286 Enamine

N'-hydroxy-3-[(piperidin-1-yl)methyl]benzene-1-carboximidamide

  • Cas Number : 1016857-30-9
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56108
Availability details for
Quantity
  • Availability
  • Add to Cart