ENA249152295 Enamine

3-cyclopropyl-6-(pyridin-3-yl)-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09741403
  • Cas Number : 954574-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56120
Availability details for
Quantity
  • Availability
  • Add to Cart